Learn More
Surmodics Inc StabilGuard™ Choice Stabilizer, 1L
SDP
Catalog No. 50949952
Encompass_Preferred
StabilGuard™ Choice Stabilizer protects the activity of proteins while simultaneously blocking nonspecific binding sites on microspheres. In microarray applications, the conformation and activity of proteins are effectively preserved in a dried state when stored with StabilGuard Choice stabilizer. The product ultimately increases signal and reduces background to improve the sensitivity of assays.
Catalog No. 50-949-952 Supplier Surmodics Inc Supplier No. SG021000
StabilGuard™ Choice Stabilizer protects the activity of proteins while simultaneously blocking nonspecific binding sites on microspheres. In microarray applications, the conformation and activity of proteins are effectively preserved in a dried state when stored with StabilGuard Choice stabilizer. The product ultimately increases signal and reduces background to improve the sensitivity of assays.