Learn More
Surmodics Inc StabilGuard™ Choice Stabilizer, 50mL
SDP
Catalog No. NC1184378
Encompass_Preferred
StabilGuard™ Choice Stabilizer protects the activity of proteins while simultaneously blocking nonspecific binding sites on microspheres. In microarray applications, the conformation and activity of proteins are effectively preserved in a dried state when stored with StabilGuard Choice stabilizer. The product ultimately increases signal and reduces background to improve the sensitivity of assays.
Catalog No. NC1184378 Supplier Surmodics Inc Supplier No. SG020050
StabilGuard™ Choice Stabilizer protects the activity of proteins while simultaneously blocking nonspecific binding sites on microspheres. In microarray applications, the conformation and activity of proteins are effectively preserved in a dried state when stored with StabilGuard Choice stabilizer. The product ultimately increases signal and reduces background to improve the sensitivity of assays.