Learn More
Surmodics Inc StabilZyme™ HRP Conjugate Stabilizer, 1L
SDP
Catalog No. 50949944
Encompass_Preferred
StabilZyme™ HRP, StabilZyme™ Protein-Free and StabilZyme™ NOBLE stabilizers provide the assay developer a complete set of tools to stabilize different HRP conjugates for long term, in-solution stability.
Catalog No. 50-949-944 Supplier Surmodics Inc Supplier No. SZ021000
StabilZyme™ HRP, StabilZyme™ Protein-Free and StabilZyme™ NOBLE stabilizers provide the assay developer a complete set of tools to stabilize different HRP conjugates for long term, in-solution stability.