Learn More
Surmodics Inc StabilZymeHRP Conjugate Stabilizer, 2L
SDP
Catalog No. 50949945
Encompass_Preferred
StabilZyme™ HRP, StabilZyme™ Protein-Free and StabilZyme™ NOBLE stabilizers provide the assay developer a complete set of tools to stabilize different HRP conjugates for long term, in-solution stability.
Catalog No. 50-949-945 Supplier Surmodics Inc Supplier No. SZ022000
StabilZyme™ HRP, StabilZyme™ Protein-Free and StabilZyme™ NOBLE stabilizers provide the assay developer a complete set of tools to stabilize different HRP conjugates for long term, in-solution stability.