Learn More
Cell Signaling Technology SignalStain™ Antibody Diluent, Immunohistochemistry, Working Solution, 100 mL
SDP
Catalog No. 50194653
Encompass
SignalStain™ Antibody Diluent is a working solution designed for use as a primary antibody diluent in immunohistochemical (IHC) assays. This reagent is formulated to improve IHC signal and background, yielding superior staining with a variety of antibodies. Please consult the specific antibody product page to confirm compatibility, as this diluent may not be suitable for use with all products recommended for immunohistochemical applications.
  • Supplied as a ready-to-use working solution.
  • Optimized to enhance staining signal and background performance in paraffin-embedded immunohistochemical assays.
  • Stable for 12 months when stored at 4°C.
  • For research use only.
Catalog No. 50-194-653 Supplier Cell Signaling Technology Supplier No. 8112L
SignalStain™ Antibody Diluent is a working solution designed for use as a primary antibody diluent in immunohistochemical (IHC) assays. This reagent is formulated to improve IHC signal and background, yielding superior staining with a variety of antibodies. Please consult the specific antibody product page to confirm compatibility, as this diluent may not be suitable for use with all products recommended for immunohistochemical applications.
  • Supplied as a ready-to-use working solution.
  • Optimized to enhance staining signal and background performance in paraffin-embedded immunohistochemical assays.
  • Stable for 12 months when stored at 4°C.
  • For research use only.