Learn More
Cell Signaling Technology Phosphate Buffered Saline with Tween 20, 20X Concentration, 1000 mL
SDP
Catalog No. 50195825
Encompass
Phosphate Buffered Saline with Tween™ 20 (PBST-20X) is a 20X concentrated buffer solution formulated for use as a wash buffer and diluent for ELISA applications. The 1X working solution contains 3.2 mM Na2HPO4, 0.5 mM KH2PO4, 1.3 mM KCl, 135 mM NaCl, and 0.05% Tween™ 20 at pH 7.4.
  • Supplied as a 20X concentrated solution
  • Yields 20 L of 1X buffer when diluted with ddH2O
  • Formulated for use in ELISA
  • Stable for 24 months when stored at room temperature
Catalog No. 50-195-825 Supplier Cell Signaling Technology Supplier No. 9809S
Phosphate Buffered Saline with Tween™ 20 (PBST-20X) is a 20X concentrated buffer solution formulated for use as a wash buffer and diluent for ELISA applications. The 1X working solution contains 3.2 mM Na2HPO4, 0.5 mM KH2PO4, 1.3 mM KCl, 135 mM NaCl, and 0.05% Tween™ 20 at pH 7.4.
  • Supplied as a 20X concentrated solution
  • Yields 20 L of 1X buffer when diluted with ddH2O
  • Formulated for use in ELISA
  • Stable for 24 months when stored at room temperature