Learn More
Cell Signaling Technology SignalStain™ Antibody Diluent, Immunohistochemical Assays, 25 mL
SDP
Catalog No. 50194654
Encompass
SignalStain™ Antibody Diluent is an immunohistochemical (IHC) buffer solution formulated to enhance signal quality and minimize background staining in various immunohistochemical applications. Supplied as a working solution, this product is optimized for the dilution of primary antibodies. Users should verify compatibility with specific antibodies by consulting the relevant IHC protocol on the antibody product page.
  • Provides superior staining results for a variety of antibodies
  • Improves IHC signal and background performance
  • Supplied as a convenient working solution
  • Stable for 12 months when stored at 4°C
Catalog No. 50-194-654 Supplier Cell Signaling Technology Supplier No. 8112S
SignalStain™ Antibody Diluent is an immunohistochemical (IHC) buffer solution formulated to enhance signal quality and minimize background staining in various immunohistochemical applications. Supplied as a working solution, this product is optimized for the dilution of primary antibodies. Users should verify compatibility with specific antibodies by consulting the relevant IHC protocol on the antibody product page.
  • Provides superior staining results for a variety of antibodies
  • Improves IHC signal and background performance
  • Supplied as a convenient working solution
  • Stable for 12 months when stored at 4°C