Learn More
Supelco Inc BUFFER 3.0 CONC TITRISOL
Catalog No. NC1729015
Encompass
(citrate/hydrochloric acid), traceable to SRM of NIST and PTB for 500 ml buffer solution, pH 3.00 ± 0.02 (20°C) Titrisol®
Catalog No. NC1729015 Supplier Supelco Inc Supplier No. 1098830001
(citrate/hydrochloric acid), traceable to SRM of NIST and PTB for 500 ml buffer solution, pH 3.00 ± 0.02 (20°C) Titrisol®