Learn More
General Laboratory Products Buffer 3, Potassium Phosphate Buffer, Antibiotic Assay, 1000 mL
Catalog No. NC2293264
Encompass_Preferred
Buffer 3 is a prepared potassium phosphate buffer designed for critical laboratory applications, specifically optimized for antibiotic assays. With a volume of 1000 mL and packaged as 12 bottles per case, it ensures stable pH conditions crucial for accurate microbiological testing. This high-quality liquid reagent adheres to USP guidelines, making it a reliable choice for sensitive analytical procedures.
Catalog No. NC2293264 Supplier General Laboratory Products Supplier No. 1000RPC70161000
Buffer 3 is a prepared potassium phosphate buffer designed for critical laboratory applications, specifically optimized for antibiotic assays. With a volume of 1000 mL and packaged as 12 bottles per case, it ensures stable pH conditions crucial for accurate microbiological testing. This high-quality liquid reagent adheres to USP guidelines, making it a reliable choice for sensitive analytical procedures.