Learn More
General Laboratory Products Buffer #4, Potassium Phosphate, 500 mL Volume
Catalog No. NC1447361
Encompass_Preferred
Buffer #4 is a potassium phosphate solution essential for antibiotic assays. Packaged in 500 mL easy-pour glass bottles, it maintains stable pH conditions for reliable lab results. Supplied in cases of 12, it supports efficient inventory management and bulk usage, distinguishing it from less effective buffering agents.
Catalog No. NC1447361 Supplier General Laboratory Products Supplier No. 500EZ7017500
Buffer #4 is a potassium phosphate solution essential for antibiotic assays. Packaged in 500 mL easy-pour glass bottles, it maintains stable pH conditions for reliable lab results. Supplied in cases of 12, it supports efficient inventory management and bulk usage, distinguishing it from less effective buffering agents.