Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell InVivoPure pH 8.0 Dilution Buffer, 50 mL

Catalog No. 501528501
Encompass_Preferred

InVivoPure™ dilution buffers are specifically formulated and tested to satisfy stringent requirements for in vivo applications. They are extremely low in endotoxin, screened for murine pathogens, and tested in animal models for toxicity.

  • Endotoxin level is <0.5 EU/mL (<0.0005 EU/μL)
  • Sterility: 0.2 μM filtered
  • Nontoxic and nonantigenic in animal models
  • Negative for a comprehensive panel of murine pathogens
  • Composition: 35 mM Na2HPO4, 1.7 mM NaH2PO4, 136 mM NaCl
  • Does not contain calcium, magnesium, phenol red, or preservatives
  • Store at 4°C; do not freeze
  • Keep contents sterile; open only in a biological safety cabinet

Catalog No. 50-152-8501 Supplier Bio X Cell Supplier No. IP0080R050ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

InVivoPure™ dilution buffers are specifically formulated and tested to satisfy stringent requirements for in vivo applications. They are extremely low in endotoxin, screened for murine pathogens, and tested in animal models for toxicity.

  • Endotoxin level is <0.5 EU/mL (<0.0005 EU/μL)
  • Sterility: 0.2 μM filtered
  • Nontoxic and nonantigenic in animal models
  • Negative for a comprehensive panel of murine pathogens
  • Composition: 35 mM Na2HPO4, 1.7 mM NaH2PO4, 136 mM NaCl
  • Does not contain calcium, magnesium, phenol red, or preservatives
  • Store at 4°C; do not freeze
  • Keep contents sterile; open only in a biological safety cabinet

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.