Learn More
Macherey-Nagel Lysis Buffer A2 (with LyseControl) 100 mL
Catalog No. NC3308453
Encompass_Preferred
Lysis Buffer A2 (with LyseControl) to be used with NucleoSpin Plasmid or NucleoSpin Plasmid (NoLid) kits
Catalog No. NC3308453 Supplier Macherey-Nagel Supplier No. 740328.100
Lysis Buffer A2 (with LyseControl) to be used with NucleoSpin Plasmid or NucleoSpin Plasmid (NoLid) kits