Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Assortment Pack - pH 4, 7, and 10, Ricca Chemical
SDP

Catalog No. 148016 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
3 x 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1480-16 3 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 1480-16 Supplier Ricca Chemical Company Supplier No. 148016
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Assortment pack contains pH 4, 7, and 10 reference buffers for routine laboratory calibration needs.

Specifications

CAS Min % 97.0, 0.33, 0.29, 0.17, 0.16, 0.13
CAS Max % 100.0, 0.34, 0.3, 0.18, 0.17, 0.14
CAS 877-24-7, 7558-79-4, 497-19-8, 7778-77-0, 144-55-8
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 4, 7, 10
Physical Form Liquid
Quantity 3 x 500 mL
Solution Type pH Buffer
Packaging Natural Poly Bottle
Grade Not Available
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.