Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Buffer Calibration Kit, Aldon
GREENER_CHOICESCI_ED

Catalog No. S20649 Shop All Aldon Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S20649 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S20649 Supplier Innovating Science™ Supplier No. IS5050
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Innovating Science pH Buffer Calibration Kit includes 500 mL of each buffer solution pH 7, pH 4, and pH 10. It also includes 120 mL of electrode storage solution.

Kit Includes:

  • 500 mL Buffer, pH 4
  • 500 mL Buffer, pH 7
  • 500 mL Buffer, pH 10
  • 120 mL Potassium Chloride, 1M

Specifications

Chemical Name or Material Buffer Calibration Kit
Concentration 1 M
For Use With (Application) pH Measurement
For Use With (Equipment) pH Probe
pH 4, 7, 10
Physical Form Liquid
Quantity 1 Kit
Solution Type pH Buffer
Molecular Weight (g/mol) Mixture
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.