Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Concentrate, 5x, pH 4.00 ± 0.01 at 25°C, Ricca Chemical
SDP

Catalog No. R1500500500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R1500500500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R1500500500 Supplier Ricca Chemical Company Supplier No. R1500500500A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

To be diluted with 4 parts Water before use

Specifications

CAS 7732-18-5, 877-24-7
CAS Max % 94.9, 5.11
Color Colorless Liquid
Concentration 5X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 4
Physical Form Liquid
Quantity 500 mL
Solution Type pH Buffer
Packaging Natural Poly Bottle
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.