Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer concentrate, pH 7.00 ± 0.02 (20°C), Supelco

Catalog No. 11100255 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-255 1 Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-255 Supplier MilliporeSigma Supelco Supplier No. 1.09887.0050
Add to Cart
Add to Cart

Traceable to SRM of NIST and PTB for 500 mL buffer solution, Titrisol™

  • Traceable to primary standard reference materials from NIST and PTB
  • Analytical standard Grade
  • Uncertainty data and traceability are documented in a comprehensive and batch-specific Certificate of Analysis.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) pH Calibration
For Use With (Equipment) pH Instruments and Electrodes
pH 7
Physical Form Aqueous Solution
Quantity 1 Bottle
Solution Type pH Buffer
Grade Analytical
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.