Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer concentrate pH 7.00, For 500 ml buffer solution, potassium dihydrogen phosphate / disodium hydrogen phosphate, Solstice

Catalog No. 60046739 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-046-739 6 x 1 Ea.
60-026-90 1 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-739 Supplier Solstice Supplier No. 387466X1EA
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

for 500 ml buffer solution, potassium dihydrogen phosphate / disodium hydrogen phosphate

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer concentrate pH 7.00
Name Note For 500 ml buffer solution, potassium dihydrogen phosphate / disodium hydrogen phosphate
CAS Min % ≥90.0000%, ≥1.0000%
CAS Max % ≤100.0000%, ≤10.0000%
CAS 7558-79-4, 7778-77-0
For Use With (Application) pH Calibration
pH 7
Physical Form Liquid
Quantity 6 x 1 Ea.
Solution Type Buffer
Packaging Ampoule
UN Number NONH for all modes of transport
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.