Learn More
Buffer concentrate pH 9.00, Buffer concentrate pH 9.00, Solstice
Shop All Solstice Research Chemicals ProductsDescription
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Chemical Name or Material | Buffer concentrate pH 9.00 |
| Name Note | Buffer concentrate pH 9.00 |
| CAS Min % | ≥92.0000%, ≥0.2000% |
| CAS | 7647-01-0, 1330-43-4 |
| CAS Max % | <5.0000% |
| Quantity | 6 x 1 Ea. |
| Solution Type | Buffer |
| Packaging | Ampoule |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.