Learn More
Ethos Biosciences Dilurex 10X PBS Buffer, 100ml, Ethos
Catalog No. 5000574200
Encompass_Preferred
Dilurex 10X PBS is a concentrated solution of phosphate buffered saline intended as a stock solution. Save space by keeping 10x concentrate and dilute to 1x pH 7.4 working concentration when needed by mixing 1 part 10X PBS with 9 parts distilled water. Skip the tedium of weighing powders and stocking raw materials that can expire. Our PBS is convenient, ready to use, and will save you hours in the lab, letting you focus on breakthroughs rather than buffer prep. Thousands of researchers trust Ethos Biosciences 10X PBS for its proven track record in top-tier publications and protocols - upgrade your productivity today.
Catalog No. 50-005-74200 Supplier Ethos Biosciences Supplier No. 4Z0004100
Dilurex 10X PBS is a concentrated solution of phosphate buffered saline intended as a stock solution. Save space by keeping 10x concentrate and dilute to 1x pH 7.4 working concentration when needed by mixing 1 part 10X PBS with 9 parts distilled water. Skip the tedium of weighing powders and stocking raw materials that can expire. Our PBS is convenient, ready to use, and will save you hours in the lab, letting you focus on breakthroughs rather than buffer prep. Thousands of researchers trust Ethos Biosciences 10X PBS for its proven track record in top-tier publications and protocols - upgrade your productivity today.