Learn More
Omega Biotek Inc DNA Wash Buffer, 100 mL
Catalog No. NC1774524
Encompass
Low Salt Spin Column Wash Solution. 100 mL concentrate makes 500 mL solution.
Catalog No. NC1774524 Supplier Omega Biotek Inc Supplier No. PS010
Low Salt Spin Column Wash Solution. 100 mL concentrate makes 500 mL solution.