Learn More
Takara Bio Buffer ENDO-EF, 1000 mL
SDP
Catalog No. NC1525527
Encompass
NucleoBond EF Buffers for endotoxin-free plasmid isolation with anion exchange columns.
Catalog No. NC1525527 Supplier Takara Bio Supplier No. 740391.1000
NucleoBond EF Buffers for endotoxin-free plasmid isolation with anion exchange columns.