Learn More
Nanotemper Technologies Inc Buffer Exploration Kit
Catalog No. NC1801707
Encompass
Your mission is to find the right buffer conditions for biophysical interaction assays, protein stability, purification, and storage. This kit will help you with your MST and TRIC assay development. Start your buffer exploration with this 96 deep-well plate containing 1 mL of each buffer condition per well.
Catalog No. NC1801707 Supplier Nanotemper Technologies Inc Supplier No. NTB001
Your mission is to find the right buffer conditions for biophysical interaction assays, protein stability, purification, and storage. This kit will help you with your MST and TRIC assay development. Start your buffer exploration with this 96 deep-well plate containing 1 mL of each buffer condition per well.