Learn More
Ethos Biosciences Hematology Buffer, pH 6.8, 16oz, Ethos
Catalog No. 5000574288
Encompass_Preferred
Hematology Buffer pH 6.8 is a buffered solution designed for hematology staining procedures. Commonly used with Romanowsky-type stains, provides the optimal pH environment for proper dye interaction and accurate cellular differentiation. The solution helps ensure consistent staining quality, clear nuclear and cytoplasmic detail, and reliable microscopic evaluation. Ideal for peripheral blood smears and bone marrow preparations, it supports routine diagnostic and research workflows.
Catalog No. 50-005-74288 Supplier Ethos Biosciences Supplier No. 554816
Hematology Buffer pH 6.8 is a buffered solution designed for hematology staining procedures. Commonly used with Romanowsky-type stains, provides the optimal pH environment for proper dye interaction and accurate cellular differentiation. The solution helps ensure consistent staining quality, clear nuclear and cytoplasmic detail, and reliable microscopic evaluation. Ideal for peripheral blood smears and bone marrow preparations, it supports routine diagnostic and research workflows.