Learn More
Ethos Biosciences Dilurex HepCas+ Buffer, 1000ml, Ethos
Catalog No. 5000574347
Encompass_Preferred
Dilurex HepCas+ Buffer is a HEPES buffer system optimized for a biocompatible pH range of 7.2-7.4 with an isotonic (0.154M) ionic stregth keeping proteins stable. It contains Casein as a carrier protein that is specially formulated to reduce non-specific binding and protease activity. Try this if you have non-specific binding using an Albumin based buffer system. Match with Casein blocking buffer for optimal performance. Formulated to optimize ELISA assays with decades of stabiilty.
Catalog No. 50-005-74347 Supplier Ethos Biosciences Supplier No. 4Z02171000
Dilurex HepCas+ Buffer is a HEPES buffer system optimized for a biocompatible pH range of 7.2-7.4 with an isotonic (0.154M) ionic stregth keeping proteins stable. It contains Casein as a carrier protein that is specially formulated to reduce non-specific binding and protease activity. Try this if you have non-specific binding using an Albumin based buffer system. Match with Casein blocking buffer for optimal performance. Formulated to optimize ELISA assays with decades of stabiilty.