Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer No-Borate, METTLER TOLEDO

Catalog No. 01913189 Shop All METTLER TOLEDO Products
Change view
Click to view available options
pH:
9.00
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH
01-913-189 9.00
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-913-189 Supplier METTLER TOLEDO Supplier No. 31049547
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The No-Borate buffer solution with pH 9.00 is a specially formulated calibration liquid that avoids borate interference, ensuring accurate pH measurements in alkaline environments.

  • Ideal for sensitive lab and industrial use, its 250 mL size supports multiple calibrations and helps maintain pH meter accuracy for quality control and compliance.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 9.00
Physical Form Solution
Quantity 250 mL
Solution Type Buffer
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.