Learn More
Cole-Parmer Oakton Traceable One-Shot™ Buffer Solution, Clear, pH 7.416; 6 x 100 mL Vials
Catalog No. 501961731 Shop All Cole-Parmer Products
Encompass_Preferred
All Oakton Traceable pH buffer solutions are NIST-traceable reference materials. Convenient, single-use vials eliminate concerns of cross-contamination; each pack contains six polyethylene vials with 100-mL of pH buffer per vial. Supplied with calibration report from an ISO 17034 and ISO 17025 accredited laboratory. Individually packed in foil pouch to prevent UV exposure that may degrade accuracy. Buffer solution lets you calibrate pH instruments to ensure accuracy If unopened, shelf life is 2 years.
Catalog No. 50-196-1731 Supplier Cole-Parmer Supplier No. 0065536
All Oakton Traceable pH buffer solutions are NIST-traceable reference materials. Convenient, single-use vials eliminate concerns of cross-contamination; each pack contains six polyethylene vials with 100-mL of pH buffer per vial. Supplied with calibration report from an ISO 17034 and ISO 17025 accredited laboratory. Individually packed in foil pouch to prevent UV exposure that may degrade accuracy. Buffer solution lets you calibrate pH instruments to ensure accuracy If unopened, shelf life is 2 years.