Learn More
Zymo Research Corporation Buffer P2 (Green), 20ml
SDP
Catalog No. 502037040
Encompass_Preferred
Buffer P2 (Green) is designed to be used with our ZR Plasmid Miniprep - Classic kit, which is used to efficiently isolate ultra-pure plasmid DNA from E. coli using Zymo-Spin column purification technology.
Catalog No. 50-203-7040 Supplier Zymo Research Corporation Supplier No. D4027220
Buffer P2 (Green) is designed to be used with our ZR Plasmid Miniprep - Classic kit, which is used to efficiently isolate ultra-pure plasmid DNA from E. coli using Zymo-Spin column purification technology.