Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer, pH 1.00, USP Reagent, Ricca Chemical
SDP

Catalog No. R14892001A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R14892001A 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R14892001A Supplier Ricca Chemical Company Supplier No. R14892001A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Buffer, pH 1.00
Name Note PH 1.00
CAS 7447-40-7, 7647-01-0, 7732-18-5
Color Colorless
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 1
Physical Form Liquid
Quantity 1 L
Solution Type pH Buffer
Odor Odorless
Packaging Natural Poly Bottle
Solubility Information Solubility in Water: Infinite
Grade USP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.