Learn More
Aqua Solutions Buffer Solution, pH 10.00 ± 0.01 @ 25°C NIST Traceable Solution (Blue) (20L)
SDP
Catalog No. NC9932955
Encompass_Preferred
Buffer Solution, pH 10.00 ± 0.01 @ 25°C NIST Traceable Solution (Blue) (20L)
Catalog No. NC9932955 Supplier Aqua Solutions Supplier No. 152520L
Buffer Solution, pH 10.00 ± 0.01 @ 25°C NIST Traceable Solution (Blue) (20L)