Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Buffer Solution, pH 10.00 ± 0.01 @ 25°C NIST Traceable Solution (Blue) (4L)
SDP

Supplier:  Aqua Solutions 15254L

Encompass_Preferred

Buffer Solution, pH 10.00 ± 0.01 @ 25°C NIST Traceable Solution (Blue) (4L)

Catalog No. NC9354069


May include imposed supplier surcharges.
Add to Cart

Description

Description

Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.