Learn More
Aqua Solutions Buffer Solution, pH 10.00 ± 0.01 @ 25°C NIST Traceable Solution (Blue) (4L)
SDP
Catalog No. NC9800048
Encompass_Preferred
Buffer Solution, pH 10.00 ± 0.01 @ 25°C NIST Traceable Solution (Blue) (4L)
Catalog No. NC9800048 Supplier Aqua Solutions Supplier No. 15254LC
Buffer Solution, pH 10.00 ± 0.01 @ 25°C NIST Traceable Solution (Blue) (4L)