Learn More
Andwin Scientific BUFFER PH 12 500ML
SDP
Catalog No. NC9731461
Encompass_Preferred
The BDH pH reference standard buffer is a clear, colorless, odorless, liquid solution. This contains potassium chloride, sodium hydroxide, and deionized water. It is classified to have a 12.00 pH at 25.0 C. This product is packaged and sold in a 500 mL poly bottle.
Catalog No. NC9731461 Supplier Andwin Scientific Supplier No. BDH5086500
The BDH pH reference standard buffer is a clear, colorless, odorless, liquid solution. This contains potassium chloride, sodium hydroxide, and deionized water. It is classified to have a 12.00 pH at 25.0 C. This product is packaged and sold in a 500 mL poly bottle.