Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer pH 2.0, MilliporeSigma™

Catalog No. MBX16521 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MBX16521 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MBX16521 Supplier MilliporeSigma Supplier No. BX16521
This item is not returnable. View return policy

Buffer pH 2.0; Clear, colorless liquid; EMD; 500mL; Poly bottle

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer pH 2.0
Color Clear
pH 2
Physical Form Liquid
Quantity 500 mL
Solution Type Buffer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.