Learn More
METTLER TOLEDO 3P INDUSTRIAL Technical Buffer Solution pH 2.00, 6 x 250 mL
Catalog No. NC1482826
Encompass_Preferred
METTLER TOLEDO certified pH buffer solutions are traceable to NIST primary standards and come with a quality inspection certificate, which guarantees the stated values and traceability. We offer convenient package sizes from single-use 20 mL pouches to 20 L Cubitainers.
Catalog No. NC1482826 Supplier METTLER TOLEDO 3P INDUSTRIAL Supplier No. 51350016
METTLER TOLEDO certified pH buffer solutions are traceable to NIST primary standards and come with a quality inspection certificate, which guarantees the stated values and traceability. We offer convenient package sizes from single-use 20 mL pouches to 20 L Cubitainers.