Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Buffer pH 4.01 ± 0.01 @ 25°C NIST Traceable (Red) (10L)
SDP

Catalog No. NC9786558
Encompass_Preferred

Buffer pH 4.01 ± 0.01 @ 25°C NIST Traceable (Red) (10L)

Catalog No. NC9786558 Supplier Aqua Solutions Supplier No. 146010L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Buffer pH 4.01 ± 0.01 @ 25°C NIST Traceable (Red) (10L)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.