Learn More
Description
- Measurement results depend on accurate calibrations, and accurate calibrations come from high quality solutions.
- Each standard solution comes with a certificate of analysis, providing traceability to NIST standards.
Specifications
Specifications
| Melting Point | 0°C |
| Boiling Point | 100°C |
| Color | Colorless |
| Description | pH Buffer |
| For Use With (Application) | Calibration |
| For Use With (Equipment) | pH Meter and Electrode |
| pH | 5 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Solution Type | Calibration |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.