Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer, Reference Standard, pH 5.80 ± 0.01 at 25°C, Ricca Chemical
SDP

Catalog No. R150900020F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R150900020F 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R150900020F Supplier Ricca Chemical Company Supplier No. R150900020F
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Provides a reliable reference standard for pH calibration in laboratory applications.

Specifications

Chemical Name or Material Buffer
Name Note pH 5.80± 0.01 at 25°C
CAS Min % 97.0, 0.82, 0.07
Color Colorless
CAS Max % 100.0, 0.86, 0.07
CAS 7778-77-0, 7558-79-4
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 5.8
Physical Form Liquid
Quantity 20 L
Solution Type pH Buffer
Packaging Cubitainer
Grade ACS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.