Learn More
Aqua Solutions Buffer pH 7.00 ± 0.01 @ 25°C NIST Traceable (Yellow) (4L)
SDP
Catalog No. NC9800047
Encompass_Preferred
Buffer pH 7.00 ± 0.01 @ 25°C NIST Traceable (Yellow) (4L)
Catalog No. NC9800047 Supplier Aqua Solutions Supplier No. 15004LC
Buffer pH 7.00 ± 0.01 @ 25°C NIST Traceable (Yellow) (4L)