Learn More
Cole-Parmer Oakton Traceable pH Standard Buffer with Calibration, Clear, pH 7, 4 L
Catalog No. NC0286269 Shop All Cole-Parmer Products
Encompass_Preferred
Oakton pH buffers are compared and traceable to NIST Standard Reference Materials. NIST-traceable reference materials include a calibration report from an ISO 17034 and ISO 17025 accredited laboratory. Calibrate at pH 7 with color-coded and colorless buffers available in a 4 L (128 oz.) bottle.
Catalog No. NC0286269 Supplier Cole-Parmer Supplier No. 0065172
Oakton pH buffers are compared and traceable to NIST Standard Reference Materials. NIST-traceable reference materials include a calibration report from an ISO 17034 and ISO 17025 accredited laboratory. Calibrate at pH 7 with color-coded and colorless buffers available in a 4 L (128 oz.) bottle.