Learn More
Cole-Parmer Oakton Traceable pH Standard Buffer with Calibration, Yellow, pH 7; 4 L
Catalog No. NC1569488 Shop All Cole-Parmer Products
Encompass_Preferred
NIST-traceable reference materials include a calibration report from an ISO 17034 and ISO 17025 accredited laboratory
Catalog No. NC1569488 Supplier Cole-Parmer Supplier No. 0065178
NIST-traceable reference materials include a calibration report from an ISO 17034 and ISO 17025 accredited laboratory