Learn More
Aqua Solutions BUFFER PH 8.0 CLEAR 1L
SDP
Catalog No. NC0951306
Encompass_Preferred
Buffer Reference Solution pH 8.00 0.02 @ 25C N.I.S.T. Traceable (1 liter)
Catalog No. NC0951306 Supplier Aqua Solutions Supplier No. 15101L
Buffer Reference Solution pH 8.00 0.02 @ 25C N.I.S.T. Traceable (1 liter)