Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer pH 8.00, 500 mL, Mettler Toledo™

Catalog No. 01914034 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-914-034 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-914-034 Supplier METTLER TOLEDO Supplier No. 30258827
Add to Cart
Edge
Add to Cart

Mettler Toledo solutions for electrode calibration and care. Quality solutions for accurate results.

  • Measurement results depend on accurate calibrations, and accurate calibrations come from high quality solutions.
  • Each standard solution comes with a certificate of analysis, providing traceability to NIST standards.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 0°C
Boiling Point 100°C
Color Colorless
Description pH Buffer
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 8
Physical Form Liquid
Quantity 500 mL
Solution Type Calibration
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.