Learn More
SCP Science Buffer pH 10 blue, 5 L
Catalog No. NC1843943
Encompass
Our full range of NIST traceable buffers are accompanied with detailed certificated of analysis complete with certified concentrations, NIST traceability, lot number and expiry date. Our buffers are available in various sizes with convenient packaging for easier dispensing.
Catalog No. NC1843943 Supplier SCP Science Supplier No. 250210502
Our full range of NIST traceable buffers are accompanied with detailed certificated of analysis complete with certified concentrations, NIST traceability, lot number and expiry date. Our buffers are available in various sizes with convenient packaging for easier dispensing.