Learn More
Cole-Parmer Oakton Traceable pH Standard Buffer with Calibration, Blue, pH 10; 500 mL
Catalog No. 501961256 Shop All Cole-Parmer Products
Encompass_Preferred
Oakton pH buffers are compared against, and are traceable to, NIST Standard Reference Materials. Each bottle includes a NIST-traceable calibration report stating accuracy at 25C to 0.010 pH. Store at room temperature. NIST-traceable reference materials include a calibration report from an ISO 17034 and ISO 17025 accredited laboratory.
Catalog No. 50-196-1256 Supplier Cole-Parmer Supplier No. 0065110
Oakton pH buffers are compared against, and are traceable to, NIST Standard Reference Materials. Each bottle includes a NIST-traceable calibration report stating accuracy at 25C to 0.010 pH. Store at room temperature. NIST-traceable reference materials include a calibration report from an ISO 17034 and ISO 17025 accredited laboratory.