Learn More
Hach Company Buffer Powder Pillows, pH 4.01 Color-coded Red, pk/15
Supplier:  Hach Company 2226995
Encompass
pH Buffer Powder Pillows for pH electrode calibration. Each powder pillow prepares 50 mL of fresh buffer solution. Color-coded for easy identification (Red). pH is 4.01 ±0.02 at 25°C. Pack of 15 Powder Pillows
Catalog No. 50-199-5573