Learn More
Hach Company Buffer Powder PIllows, pH 4.01 & pH 7.00, pk/2x15
Supplier:  Hach Company 2299264
Encompass
pH Buffer Powder Pillows for pH electrode calibration. Each powder pillow prepares 50 mL of fresh buffer solution. Color-coded for easy identification. pH is ±0.02 at 25°C. Pack of 15 pillows of pH 4.01 (red) and 15 pillows of pH 7.00 (yellow).
Catalog No. 50-199-5754