Learn More
Hach Company Buffer Powder Pillows, pH 4.50, 50 mL, pk/25
Supplier:  Hach Company 89568
Encompass
For endpoint standardization in Alkalinity determination by visual titration. Sample size: 50 mL. Pack of 25 Powder Pillows
Catalog No. 50-199-4463