Learn More
Sigma Aldrich Fine Chemicals Biosciences pH 4 Buffer Solution | N/A | 500 mL | Clear
Catalog No. NC2596928
Encompass
BUFFER PH4 500ML CLEAR | Purity: N/A | Mol Wt: N/A | CAS No: N/A | MDL No: N/A | 500mLA pH 4 buffered solution with a volume of 500 milliliters used as a pH reference standard, for calibrating pH meters, and preparing pH 4 solutions. The clear solution enables visual inspection.
Catalog No. NC2596928 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. BX16341500ML
BUFFER PH4 500ML CLEAR | Purity: N/A | Mol Wt: N/A | CAS No: N/A | MDL No: N/A | 500mLA pH 4 buffered solution with a volume of 500 milliliters used as a pH reference standard, for calibrating pH meters, and preparing pH 4 solutions. The clear solution enables visual inspection.