Learn More
Hach Company Buffer Powder Pillows, pH 7.00 Color-coded Yellow, pk/15
Catalog No. 501995576 Shop All Hach Company Products
Encompass
pH Buffer Powder Pillows for pH electrode calibration. Each powder pillow prepares 50 mL of fresh buffer solution. Color-coded for easy identification (Yellow). pH is 7.00 ±0.02 at 25°C. Pack of 15 Powder Pillows
Catalog No. 50-199-5576 Supplier Hach Company Supplier No. 2227095
pH Buffer Powder Pillows for pH electrode calibration. Each powder pillow prepares 50 mL of fresh buffer solution. Color-coded for easy identification (Yellow). pH is 7.00 ±0.02 at 25°C. Pack of 15 Powder Pillows