Learn More
Hach Company Buffer Powder Pillows, pH 8.30, 50 mL, pk/25
Supplier:  Hach Company 89868
Encompass
Buffer for preparing monochloramine standards and detecting visual alkalinity endpoints. pH 8.30, Sample Size: 50 mL sample. Pack of 25 Powder Pillows.
Catalog No. 50-199-4464